missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-1,4-difluorobenzene, 98%, Thermo Scientific™

Product Code. 11482237
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
This item is not returnable. View return policy

Product Code. 11482237

Brand: Thermo Scientific Alfa Aesar A14178.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2367-91-1
Molecular Formula C6H3ClF2
Molecular Weight (g/mol) 148.537
MDL Number MFCD00042571
InChI Key XWCKSJOUZQHFKI-UHFFFAOYSA-N
PubChem CID 137561
IUPAC Name 2-chloro-1,4-difluorobenzene
SMILES C1=CC(=C(C=C1F)Cl)F

Specifications

Density 1.352
Boiling Point 126°C to 128°C
Flash Point 20°C (68°F)
Odor Aromatic
Refractive Index 1.479
Quantity 5 g
UN Number UN1993
Beilstein 2081082
Synonym 1-chloro-2,5-difluorobenzene, 2,5-difluorochlorobenzene, benzene,2-chloro-1,4-difluoro, benzene, 2-chloro-1,4-difluoro, pubchem8529, acmc-20ai5u, ksc493a9f, 2-chloro-1,4-difluoro-benzene, 2-chloro-4-fluoro-fluorobenzene, 2-chloranyl-1,4-bis fluoranyl benzene
Formula Weight 148.54
Percent Purity 98%
Chemical Name or Material 2-Chloro-1,4-difluorobenzene
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.