missing translation for 'onlineSavingsMsg'
Learn More

2-Chloro-4-(trifluoromethyl)benzonitrile, 98%, Thermo Scientific™

Product Code. 11421440
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11421440

Brand: Thermo Scientific Alfa Aesar H26894.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1813-33-8
Molecular Formula C8H3ClF3N
Molecular Weight (g/mol) 205.564
MDL Number MFCD00084939
InChI Key GEHMLBFNZKJDQM-UHFFFAOYSA-N
Synonym 2-chloro-4-trifluoromethyl benzonitrile, 3-chloro-4-cyanobenzotrifluoride, 3-chloro-4-cyano-benzotrifluoride, 4-trifluoromethyl-2-chlorobenzonitrile, benzonitrile, 2-chloro-4-trifluoromethyl, 2-chloro-4-trifluoromethyl benzenecarbonitrile, pubchem4745, acmc-209eho, buttpark 9757-31, ksc494o6f
PubChem CID 2736476
IUPAC Name 2-chloro-4-(trifluoromethyl)benzonitrile
SMILES C1=CC(=C(C=C1C(F)(F)F)Cl)C#N

Specifications

Melting Point 82°C to 83°C (8mmHg)
Density 1.389
Boiling Point 192°C to 193°C
Flash Point >104°C (220°F)
Odor Characteristic
Refractive Index 1.484
Quantity 5 g
UN Number UN3276
Formula Weight 205.57
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-(trifluoromethyl)benzonitrile
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.