missing translation for 'onlineSavingsMsg'
Learn More
2-Ethoxy-5-(trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific™ Product Code.: 15439895

2-Ethoxy-5-(trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific™

Product Code. 15439895
1 g
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15439895

Brand: Thermo Scientific Alfa Aesar H52541.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 850593-10-1
Molecular Formula C9H10BF3O3
Molecular Weight (g/mol) 233.981
MDL Number MFCD06801695
InChI Key IOCXELVDHLDNMQ-UHFFFAOYSA-N
Synonym 2-ethoxy-5-trifluoromethylphenylboronic acid, 2-ethoxy-5-trifluoromethylphenyl boronic acid, 2-ethoxy-5-trifluoromethyl benzeneboronic acid, 6-ethoxy-3-trifluoromethyl phenylboronic acid, 2-ethoxy-5-trifluoromethyl phenylboronic acid, 2-ethoxy-5-trifluoromethyl phenyl boronic acid, akos brn-0565, acmc-209q3k, 2-ethoxy-5-trifluoromethylphenylboronicacidShow MoreShow Less
PubChem CID 23005337
IUPAC Name [2-ethoxy-5-(trifluoromethyl)phenyl]boronic acid
SMILES B(C1=C(C=CC(=C1)C(F)(F)F)OCC)(O)O

Specifications

Melting Point 100°C to 104°C
Quantity 1 g
Formula Weight 233.98
Percent Purity 98%
Chemical Name or Material 2-Ethoxy-5-(trifluoromethyl)benzeneboronic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10169689 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Ethoxy-5-(trifluoromethyl)benzeneboronic acid, 98%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.