missing translation for 'onlineSavingsMsg'
Learn More

2-Fluoro-5-(trifluoromethoxy)phenol, 97%, Thermo Scientific™

86.50 € - 284.00 €

Chemical Identifiers

CAS 886498-03-9
Molecular Formula C7H4F4O2
Molecular Weight (g/mol) 196.101
MDL Number MFCD04115893
InChI Key MKRRYWGEQVWJCA-UHFFFAOYSA-N
Synonym 2-fluoro-5-trifluoromethoxy phenol, acmc-209qvx, phenol, 2-fluoro-5-trifluoromethoxy
PubChem CID 3790875
IUPAC Name 2-fluoro-5-(trifluoromethoxy)phenol
SMILES C1=CC(=C(C=C1OC(F)(F)F)O)F
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11435950
View Documents
Thermo Scientific Alfa Aesar
H26535.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11445950
View Documents
Thermo Scientific Alfa Aesar
H26535.06
5 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications

Chemical Identifiers

886498-03-9
196.101
MKRRYWGEQVWJCA-UHFFFAOYSA-N
3790875
C1=CC(=C(C=C1OC(F)(F)F)O)F
C7H4F4O2
MFCD04115893
2-fluoro-5-trifluoromethoxy phenol, acmc-209qvx, phenol, 2-fluoro-5-trifluoromethoxy
2-fluoro-5-(trifluoromethoxy)phenol

Specifications

886498-03-9
MFCD04115893
MKRRYWGEQVWJCA-UHFFFAOYSA-N
2-fluoro-5-(trifluoromethoxy)phenol
3790875
97%
C7H4F4O2
2-fluoro-5-trifluoromethoxy phenol, acmc-209qvx, phenol, 2-fluoro-5-trifluoromethoxy
C1=CC(=C(C=C1OC(F)(F)F)O)F
196.101
196.1
2-Fluoro-5-(trifluoromethoxy)phenol
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H315-H319-H335
Toxic if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302-H315-H319-H335

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Product Certifications