missing translation for 'onlineSavingsMsg'
Learn More

2-(Methylamino)ethanol, 99%

Product Code. p-3736887 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
500 g
2.5 kg
10 kg
Packaging:
Glass bottle
Metal Drum
Unit Size:
10kg
2.5kg
500g
This item is not returnable. View return policy

Product Code. 10522751

Brand: Thermo Scientific Acros 158265000

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 109-83-1 | C3H9NO | 75.11 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-83-1
Molecular Formula C3H9NO
Molecular Weight (g/mol) 75.11
MDL Number MFCD00002839
InChI Key OPKOKAMJFNKNAS-UHFFFAOYSA-N
Synonym 2-methylamino ethanol, n-methylethanolamine, methylethanolamine, ethanol, 2-methylamino, methylethylolamine, n-methylaminoethanol, n-methylmonoethanolamine, n-monomethylethanolamine, 2-hydroxyethyl methylamine, monomethylaminoethanol
PubChem CID 8016
ChEBI CHEBI:21763
IUPAC Name 2-(methylamino)ethanol
SMILES CNCCO

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point -3.0°C
Color Colorless to Yellow
Density 0.9300g/mL
Boiling Point 159.0°C
Flash Point 72°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Linear Formula CH3NHCH2CH2OH
Refractive Index 1.4380 to 1.4400
Quantity 500 g
Beilstein 04, 276
Merck Index 15, 6089
Specific Gravity 0.93
Solubility Information Solubility in water: miscible
Viscosity 13 mPa.s (20°C)
Formula Weight 75.11
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material 2-(Methylamino)ethanol
Show More Show Less
compliance-icons compliance-icons compliance-icons
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 1
  • Skin corrosion/irritation Category 1 B
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H302-Harmful if swallowed.
  • H312-Harmful in contact with skin.
  • H314-Causes severe skin burns and eye damage.
  • H318-Causes serious eye damage.
  • H335-May cause respiratory irritation.
  • H373-May cause damage to organs.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.