Learn More
Greiner Bio-One 2 mL Sterile Serological Pipets
GREENER_CHOICE
Drip-free pipetting design with high optical clarity and maximum accuracy.
Brand:  Greiner Bio-One 710160
 View more versions of this product
Product Code. 10553144
  • 171.43 € / Pack of 1000
In Stock
to see stock.
Edge
Explore more special offers
Add to basket
Description
Description
  • High-grade polystyrene
  • Clear, easily legible graduation
  • Colored vertical stripe for ease in reading the volume
  • With synthetic fibrous filter material for protection against
    TRUSTED_SUSTAINABILITY
Specifications
Specifications
Shape: Standard
Sterile
1/100
2 mL
Polystyrene
Yes
Individual Plastic Wrap
Serological Pipet
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Greiner Bio-One 2 mL Sterile Serological Pipets > Individually wrapped; Packaging: Plastic/Plastic