missing translation for 'onlineSavingsMsg'
Learn More
2-Nitro-m-xylene, 99%, Thermo Scientific Chemicals Product Code.: 11307495

2-Nitro-m-xylene, 99%, Thermo Scientific Chemicals

Product Code. 11307495
500 g
Click to view available options
Quantity:
100 g
500 g
This item is not returnable. View return policy

Product Code. 11307495

Brand: Thermo Scientific Alfa Aesar L05546.36

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

2-Nitro-m-xylene is employed as a pharmaceutical intermediate in organic synthesis. It is also used in the preparation of Direct Violet 7 and solvent Red 26. Further, it is used in the petrochemical industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
2-Nitro-m-xylene is employed as a pharmaceutical intermediate in organic synthesis. It is also used in the preparation of Direct Violet 7 and solvent Red 26. Further, it is used in the petrochemical industry.

Solubility
Miscible with ethanol. Immiscible with water.

Notes
Incompatible with strong oxidizing agents and strong bases.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 81-20-9
Molecular Formula C8H9NO2
Molecular Weight (g/mol) 151.165
MDL Number MFCD00007181
InChI Key HDFQKJQEWGVKCQ-UHFFFAOYSA-N
Synonym 2-nitro-m-xylene, 2,6-dimethylnitrobenzene, benzene, 1,3-dimethyl-2-nitro, 2,6-dimethyl-1-nitrobenzene, m-xylene, 2-nitro, ccris 3119, 1,3-dimethyl-2-nitro-benzene, dsstox_cid_5134, dsstox_rid_77678, dsstox_gsid_25134Show MoreShow Less
PubChem CID 6672
IUPAC Name 1,3-dimethyl-2-nitrobenzene
SMILES CC1=C(C(=CC=C1)C)[N+](=O)[O-]

Specifications

Melting Point 14°C to 16°C
Density 1.11
Boiling Point 225°C to 227°C
Flash Point 87°C (188°F)
Refractive Index 1.522
Quantity 500 g
UN Number UN1665
Beilstein 2046066
Solubility Information Miscible with ethanol. Immiscible with water.
Formula Weight 151.17
Percent Purity 99%
Chemical Name or Material 2-Nitro-m-xylene
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249034 Certificate TypeCertificate of Analysis Date21/12/2023 Catalog NumberL05546.36,L05546.22
Lot Number10248767 Certificate TypeCertificate of Analysis Date05/12/2023 Catalog NumberL05546.36,L05546.22
Lot Number10244760 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberL05546.36,L05546.22
Lot Number10245719 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberL05546.36,L05546.22
Lot Number10245727 Certificate TypeCertificate of Analysis Date09/05/2023 Catalog NumberL05546.36

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Nitro-m-xylene, 99%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.