Learn More
2-Nitrotoluene, 99+%, Thermo Scientific Chemicals Product Code.: 11475503
2-Nitrotoluene, 99+%, Thermo Scientific Chemicals
Product Code. 11475503
500 g
Click to view available options
Quantity:
100 g
500 g
2500 g
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Product Code. 11475503
Brand: Thermo Scientific Alfa Aesar A13446.36
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
Regulatory: This product is an isolated intermediate transported in accordance with Regulation (EC) 1907/2006 (REACH). All customers must complete a declaration before delivery.
Used as an intermediate, reagent, as a precursor and as a nitrogen, carbon, energy supplement
2-Nitrotoluene is used as an intermediate for the o-toludine synthesis, which finds application in the dyestuff industry. It is employed as a reagent in detecting and photometric determining oxidizing agent. It is also used in the determination of prussic acid content in the air. It acts as a precursor to prepare 2-amino-4-chlorotoluene, 2-amino-6-chlorotoluene, nitrotoluenesulfonic acids and toluene nitrosulfonic chlorides. It acts as a nitrogen supplement in the culture medium of pseudomonas sp. strain ClS1. It acts as a carbon and energy supplement in the culture medium of Pseudomonas sp. strain JS42.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Nitrotoluene is used as an intermediate for the o-toludine synthesis, which finds application in the dyestuff industry. It is employed as a reagent in detecting and photometric determining oxidizing agent. It is also used in the determination of prussic acid content in the air. It acts as a precursor to prepare 2-amino-4-chlorotoluene, 2-amino-6-chlorotoluene, nitrotoluenesulfonic acids and toluene nitrosulfonic chlorides. It acts as a nitrogen supplement in the culture medium of pseudomonas sp. strain ClS1. It acts as a carbon and energy supplement in the culture medium of Pseudomonas sp. strain JS42.
Solubility
Miscible with acetone, benzene, chloroform, diethyl ether, ethanol and petroleum ether. Immiscible with water.
Notes
Store in a cool place. Incompatible with oxidizing agents, strong bases, sulfuric acid, reducing agents, hydrogen and sodium.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 88-72-2
Molecular Formula C7H7NO2
Molecular Weight (g/mol) 137.138
MDL Number MFCD00007157
InChI Key PLAZTCDQAHEYBI-UHFFFAOYSA-N
Synonym 2-nitrotoluene, o-nitrotoluene, o-nitrotoluol, o-methylnitrobenzene, benzene, 1-methyl-2-nitro, toluene, o-nitro, 2-methyl-1-nitrobenzene, 2-nitrotoluol, ortho-nitrotoluol, 2-methylnitrobenzeneShow MoreShow Less
PubChem CID 6944
ChEBI CHEBI:33098
IUPAC Name 1-methyl-2-nitrobenzene
SMILES CC1=CC=CC=C1[N+](=O)[O-]
Specifications
Melting Point -4°C
Density 1.166
Boiling Point 224°C to 226°C
Flash Point 106°C (222°F)
Refractive Index 1.545
Quantity 500 g
UN Number UN1664
Beilstein 1907580
Merck Index 14,6650
Solubility Information Miscible with acetone,benzene,chloroform,diethyl ether,ethanol and petroleum ether; Immiscible with water.
Formula Weight 137.14
Percent Purity ≥99%
Chemical Name or Material 2-Nitrotoluene
Show More Show Less
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10219428 Certificate TypeCertificate of Analysis Date29/12/2023 Catalog NumberA13446.22,A13446.36,A13446.0E
Lot Number10244720 Certificate TypeCertificate of Analysis Date24/10/2023 Catalog NumberA13446.22,A13446.36,A13446.0E
Lot Number10233000 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA13446.22,A13446.36,A13446.0E
Lot Number10209103 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13446.22,A13446.36,A13446.0E
Lot Number10157488 Certificate TypeCertificate of Analysis Date07/08/2020 Catalog NumberA13446.22,A13446.36,A13446.0E
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
2-Nitrotoluene, 99+%, Thermo Scientific Chemicals >