missing translation for 'onlineSavingsMsg'
Learn More
2-(Tetrahydropyran-4-yloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 97%, Thermo Scientific™ Product Code.: 10193374

2-(Tetrahydropyran-4-yloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 97%, Thermo Scientific™

Product Code. 10193374
1 g, Amber Glass Bottle
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
This item is not returnable. View return policy

Product Code. 10193374

Brand: Thermo Scientific Maybridge CC58339DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 910036-98-5
Molecular Formula C16H24BNO4
Molecular Weight (g/mol) 305.18
MDL Number MFCD09702390
InChI Key KGCJDMWHIVJQGF-UHFFFAOYSA-N
Synonym 2-tetrahydro-pyran-4-yloxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl-pyridine, 2-tetrahydropyran-4-yloxy-5-4,4,5,5-tetramethyl-1,3,2 dioxaborolan-2-yl pyridine, 2-tetrahydro-2h-pyran-4-yl oxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-oxan-4-yloxy-5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-tetrahydro-2h-pyran-4-yloxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 2-oxan-4-yloxy-5-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pubchem20144, 2-tetrahydropyran-4-yloxy pyridine-5-boronic acid pinacol ester, 6-tetrahydropyran-4-yloxy pyridine-3-boronic acid pinacol ester, 6-tetrahydro-2h-pyran-4-yl oxy pyridin-3-yl boronic acid pinacol ester
PubChem CID 24229583
IUPAC Name 2-(oxan-4-yloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
SMILES CC1(C)OB(OC1(C)C)C1=CN=C(OC2CCOCC2)C=C1

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 305.18
Percent Purity 97%
Chemical Name or Material 2-(Tetrahydropyran-4-yloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-(Tetrahydropyran-4-yloxy)-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.