missing translation for 'onlineSavingsMsg'
Learn More
2-Trifluoromethyl-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine hydrochloride, 97%, Thermo Scientific™ Product Code.: 15435165

2-Trifluoromethyl-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine hydrochloride, 97%, Thermo Scientific™

Product Code. 15435165
250 mg
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
This item is not returnable. View return policy

Product Code. 15435165

Brand: Thermo Scientific Alfa Aesar H33501.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 911064-58-9
Molecular Formula C7H9ClF3N3
Molecular Weight (g/mol) 227.62
MDL Number MFCD09999262
InChI Key WQJNJYFZTOYWPU-UHFFFAOYSA-N
Synonym 2-trifluoromethyl-5,6,7,8-tetrahydroimidazo 1,2-a pyrazine hydrochloride, 2-trifluoromethyl-5,6,7,8-tetrahydroimidazo 1,2-a pyrazine hcl, 2-trifluoromethyl-5h,6h,7h,8h-imidazo 1,2-a pyrazine hydrochloride, 2-trifluoromethyl-5,6,7,8-tetrahydroimidazo-1,2-a pyrazine hydrochloride, 2-trifluoromethyl-5,6,7,8-tetrahydroimidazo 1,2-a pyrazine 2hcl, c7h8f3n3.clh, 2-trifluoromethyl-5,6,7,8-tetrahydro-imidazo 1,2-a pyrazine hcl salt, 2-trifluoromethyl-5,6,7,8-tetrahydroimidazo 1,2-a pyrazine-hydrogen chloride 1/1Show MoreShow Less
PubChem CID 44118283
IUPAC Name 2-(trifluoromethyl)-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine;hydrochloride
SMILES Cl.FC(F)(F)C1=CN2CCNCC2=N1

Specifications

Melting Point ∽220°C (decomposition)
Quantity 250 mg
Sensitivity Hygroscopic
Formula Weight 227.62
Percent Purity 97%
Chemical Name or Material 2-Trifluoromethyl-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine hydrochloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10174185 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
2-Trifluoromethyl-5,6,7,8-tetrahydroimidazo[1,2-a]pyrazine hydrochloride, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.