Learn More
Fisher Chemical 200LT Toluene, 99 %, extra pure, SLR
Brand:  Fisher Chemical T/2200/27
200LT Toluene, 99 %, extra pure, SLR
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 10357694
  • ONLINE EXCLUSIVE PRICE
    917.00 €
    884.00 € / 200 L
    Save 33.00 €
    3% Off