Learn More
Greiner Bio-One 25 mL Sterile Serological Pipets
GREENER_CHOICE
Drip-free pipetting design with high optical clarity and maximum accuracy.
Brand:  Greiner Bio-One 760107
 View more versions of this product
Product Code. 10666984
  • 268.00 € / Pack of 200
Estimated Shipment: 27-11-2024
to see stock.
Explore more special offers
Add to basket
Description
Description
  • High-grade polystyrene
  • Clear, easily legible graduation
  • Colored vertical stripe for ease in reading the volume
  • With synthetic fibrous filter material for protection against
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Shape: Standard
Sterile
2/10
25 mL
Polystyrene
Yes
Bulk
Serological Pipet