missing translation for 'onlineSavingsMsg'
Learn More

(2R,4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid methyl ester, 95%, 98% ee, Thermo Scientific Chemicals

249.00 € - 437.18 €

Chemical Identifiers

CAS 321744-26-7
Molecular Formula C12H21NO5
Molecular Weight (g/mol) 259.30
MDL Number MFCD02259744
InChI Key RNMVWSAJMIKMDY-DTWKUNHWSA-N
PubChem CID 2734504
IUPAC Name 1-O-tert-butyl 2-O-methyl (2R,4S)-4-hydroxypiperidine-1,2-dicarboxylate
SMILES COC(=O)[C@H]1C[C@@H](O)CCN1C(=O)OC(C)(C)C
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10170272
SDS View Documents
Thermo Scientific Acros
362975000
500 mg Glass bottle N/A N/A N/A
10028781
SDS View Documents
Thermo Scientific Acros
362970010
1 g Glass bottle
437.18 €
1g
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : TSCA

Product Identifier
  • (2R,4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid methyl ester

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
(2R,4S)-N-BOC-4-Hydroxypiperidine-2-carboxylic acid methyl ester, 95%, 98% ee, Thermo Scientific Chemicals

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.