missing translation for 'onlineSavingsMsg'
Learn More

(2S,4S)-1-Boc-4-diphenylphosphino-2-(diphenylphosphinomethyl)pyrrolidine

Product Code. 11342409
missing translation for 'orderingAttributeHoverText'
Quantity:
250 mg
1 g
missing translation for 'unitSize'
1g
250mg
This item is not returnable. View return policy

Product Code. 11342409

missing translation for 'mfr': Thermo Scientific Alfa Aesar H27321.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 61478-28-2 | C34H37NO2P2 | 553.62 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 61478-28-2
Molecular Formula C34H37NO2P2
Molecular Weight (g/mol) 553.62
MDL Number MFCD01631275
InChI Key BFMKBYZEJOQYIM-FEHSQGEHNA-N
Synonym 2s,4s---n-boc-4-diphenylphosphino-2-diphenylphosphinomethyl-pyrrolidine, bppm, s,s-bppm, 2s,4s-1-tert-butoxycarbonyl-4-diphenylphosphino-2-diphenylphosphinomethyl pyrrolidine, 2s,4s---1-boc-4-diphenylphosphino-2-diphenylphosphinomethyl pyrrolidine, 2s,4s---2-diphenylphosphinomethyl-4-diphenylphosphino-n-t-butoxycarbonyl pyrrolidine, 2s,4s-tert-butyl 4-diphenylphosphino-2-diphenylphosphino methyl pyrrolidine-1-carboxylate, tert-butyl 2s,4s-4-diphenylphosphanyl-2-diphenylphosphanyl methyl pyrrolidine-1-carboxylate, 2s,4s-1-boc-4-diphenylphosphino-2-diphenylphosphinomethyl pyrrolidine
PubChem CID 10209545
IUPAC Name tert-butyl (2S,4S)-4-diphenylphosphanyl-2-(diphenylphosphanylmethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1C[C@H](C[C@H]1CP(C1=CC=CC=C1)C1=CC=CC=C1)P(C1=CC=CC=C1)C1=CC=CC=C1

Specifications

Melting Point 100°C to 103°C
Quantity 1 g
Formula Weight 553.62
Chemical Name or Material (2S,4S)-1-Boc-4-diphenylphosphino-2-(diphenylphosphinomethyl)pyrrolidine

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.