missing translation for 'onlineSavingsMsg'
Learn More

3,4,5-Trifluorobenzenesulfonyl chloride, 98%, Thermo Scientific™

Product Code. 11379935
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 351003-43-5
Molecular Formula C6H2ClF3O2S
Molecular Weight (g/mol) 230.59
MDL Number MFCD03094559
InChI Key HSEHKULCGSMHAE-UHFFFAOYSA-N
Synonym 3,4,5-trifluorobenzene-1-sulfonyl chloride, 3,4,5-trifluorobenzenesulfonylchloride, 3,4,5-trifluorobenzenesulphonyl chloride, benzenesulfonyl chloride, 3,4,5-trifluoro, pubchem11715, acmc-1cttq, chloro 3,4,5-trifluorophenyl sulfone, 3,4,5-trifluorophenylsulfonyl chloride, benzenesulfonylchloride, 3,4,5-trifluoro, 3,4,5-trifluorobenzenesulfonyl chloride
PubChem CID 2778858
IUPAC Name 3,4,5-trifluorobenzene-1-sulfonyl chloride
SMILES FC1=CC(=CC(F)=C1F)S(Cl)(=O)=O

Specifications

Density 1.643
Boiling Point ∼230°C
Flash Point >100°C (212°F)
Refractive Index 1.497
Quantity 5 g
UN Number UN3265
Sensitivity Moisture sensitive
Formula Weight 230.59
Percent Purity 98%
Chemical Name or Material 3,4,5-Trifluorobenzenesulfonyl chloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.