missing translation for 'onlineSavingsMsg'
Learn More
3,4-Dichlorobenzotrifluoride, 97%, Thermo Scientific™ Product Code.: 11319539

3,4-Dichlorobenzotrifluoride, 97%, Thermo Scientific™

Product Code. 11319539
250 g
Click to view available options
Quantity:
10 g
50 g
250 g
500 g
This item is not returnable. View return policy

Product Code. 11319539

Brand: Thermo Scientific Alfa Aesar A15098.30

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 328-84-7
Molecular Formula C7H3Cl2F3
Molecular Weight (g/mol) 214.996
MDL Number MFCD00000555
InChI Key XILPLWOGHPSJBK-UHFFFAOYSA-N
Synonym 3,4-dichlorobenzotrifluoride, 1,2-dichloro-4-trifluoromethyl benzene, benzene, 1,2-dichloro-4-trifluoromethyl, 3,4-dichloro-alpha,alpha,alpha-trifluorotoluene, 3,4-dichlorotrifluoromethylbenzene, unii-7g60xcu4gg, 3,4-dichlorophenyltrifluoromethane, 7g60xcu4gg, 3,4-dichloro-1-trifluoromethyl benzene, 3,4-dcbteShow MoreShow Less
PubChem CID 9481
IUPAC Name 1,2-dichloro-4-(trifluoromethyl)benzene
SMILES C1=CC(=C(C=C1C(F)(F)F)Cl)Cl

Specifications

Melting Point -12°C
Density 1.479
Boiling Point 173°C to 174°C
Flash Point 65°C (149°F)
Refractive Index 1.475
Quantity 250 g
UN Number UN1760
Beilstein 1950151
Formula Weight 215
Percent Purity 97%
Chemical Name or Material 3,4-Dichlorobenzotrifluoride
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

2 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberG2772A Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberD1996A Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,4-Dichlorobenzotrifluoride, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.