missing translation for 'onlineSavingsMsg'
Learn More

3-(4-tert-Butylphenyl)propionic acid, 97%

Product Code. 15430458
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 15430458

Brand: Thermo Scientific Alfa Aesar H54011.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1208-64-6 | C13H18O2 | 206.285 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1208-64-6
Molecular Formula C13H18O2
Molecular Weight (g/mol) 206.285
MDL Number MFCD00796482
InChI Key BNJYANVQFVSYEK-UHFFFAOYSA-N
Synonym 3-4-tert-butyl-phenyl-propionic acid, 3-4-tert-butyl phenyl propanoic acid, 3-4-tert-butylphenyl propanoic acid, 4-1,1-dimethylethyl benzenepropanoic acid, 4-t-butylhydrocinnamic acid, 4-t-butylhydrocinnamic aicd, 4-tert-butylhydrocinnamic acid, 3-4-t-butylphenyl propionic acid, 3-4-t-butylphenyl propanoic acid
PubChem CID 4962173
IUPAC Name 3-(4-tert-butylphenyl)propanoic acid
SMILES CC(C)(C)C1=CC=C(C=C1)CCC(=O)O

Specifications

Melting Point 102°C
Quantity 5 g
Formula Weight 206.28
Percent Purity 97%
Chemical Name or Material 3-(4-tert-Butylphenyl)propionic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.