missing translation for 'onlineSavingsMsg'
Learn More
3,5-Dibromophenol, 97%, Thermo Scientific Chemicals Product Code.: 15407128

3,5-Dibromophenol, 97%, Thermo Scientific Chemicals

Product Code. 15407128
10 g
Click to view available options
Quantity:
2 g
10 g
This item is not returnable. View return policy

Product Code. 15407128

Brand: Thermo Scientific Alfa Aesar H60642.09

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3,5-Dibromophenol is used as a pharmaceutical intermediate. It is also used to prepare aromatic polyesters and drugs for arthritis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3,5-Dibromophenol is used as a pharmaceutical intermediate. It is also used to prepare aromatic polyesters and drugs for arthritis.

Solubility
Sightly soluble in water. Soluble in methanol.

Notes
Incompatible with strong oxidizing agents. Light sensitive.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 626-41-5
Molecular Formula C6H4Br2O
Molecular Weight (g/mol) 251.905
MDL Number MFCD06411376
InChI Key PZFMWYNHJFZBPO-UHFFFAOYSA-N
PubChem CID 12280
IUPAC Name 3,5-dibromophenol
SMILES C1=C(C=C(C=C1Br)Br)O

Specifications

Melting Point 79°C to 83°C
Quantity 10 g
UN Number UN2811
Synonym phenol, 3,5-dibromo, 3,5-dibromo-phenol, 3,5-dibromphenol, 3, 5-dibromophenol, 3-,5-dibromophenol, pubchem23831, acmc-1az7g, 3,5-dibromophenol, ksc493e8n, akos akm01584
Solubility Information Sightly soluble in water. Soluble in methanol.
Formula Weight 251.9
Percent Purity 97%
Chemical Name or Material 3,5-Dibromophenol

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberY14I016 Certificate TypeCertificate of Analysis Date29/11/2022 Catalog NumberH60642.04,H60642.09
Lot Number81302549 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60642.04,H60642.09
Lot Number5012R18A Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60642.04,H60642.09
Lot NumberU10H058 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60642.04,H60642.09
Lot Number81500015 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberH60642.04,H60642.09

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3,5-Dibromophenol, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.