Learn More
3,5-Dichlorophenylhydrazine hydrochloride, 95%
Description
3,5-Dichlorophenyl)hydrazine Hydrochloride is a building block that has been used for the synthesis of 2H-benzo[b][1,4]oxazin-3(4H)-one derivatives to be used as and anti-leukemic agent.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
3,5-Dichlorophenyl)hydrazine Hydrochloride is a building block that has been used for the synthesis of 2H-benzo[b][1,4]oxazin-3(4H)-one derivatives to be used as and anti-leukemic agent.
Solubility
Soluble in water.
Notes
Store in cool, dry place in tightly closed container. Store away from oxidizing agent.
Specifications
Specifications
| Melting Point | ∼204°C (decomposition) |
| Quantity | 1 g |
| Beilstein | 4208459 |
| Solubility Information | Soluble in water. |
| Formula Weight | 213.5 |
| Percent Purity | 97% |
| Chemical Name or Material | 3,5-Dichlorophenylhydrazine hydrochloride |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.