missing translation for 'onlineSavingsMsg'
Learn More

3-Amino-3-(2-chlorophenyl)propionic acid, 97%, Thermo Scientific™

40.50 € - 87.10 €

Chemical Identifiers

CAS 68208-20-8
Molecular Formula C9H10ClNO2
Molecular Weight (g/mol) 199.63
MDL Number MFCD01871299
InChI Key NXXFYRJVRISCCP-MRVPVSSYSA-N
Synonym 3-amino-3-2-chlorophenyl propanoic acid, 3-amino-3-2-chloro-phenyl-propionic acid, 3-2-chlorophenyl-dl-beta-alanine, 3-amino-3-2-chlorophenyl propionic acid, dl-beta-2-chlorophenyl alanine, pubchem13916, rarechem ak hc t319, 3-amino-3-2-chlorophenyl propanoicacid, benzenepropanoic acid, .beta.-amino-2-chloro, r/s-3-amino-3-2-chloro-phenyl-propionic acid
PubChem CID 239929
IUPAC Name 3-amino-3-(2-chlorophenyl)propanoic acid
SMILES [NH3+][C@H](CC([O-])=O)C1=CC=CC=C1Cl
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11309765
SDS View Documents
Thermo Scientific Alfa Aesar
L19796.MD
250 mg This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11399755
SDS View Documents
Thermo Scientific Alfa Aesar
L19796.03
1 g This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

missing translation for 'tsca' : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Amino-3-(2-chlorophenyl)propionic acid, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.