missing translation for 'onlineSavingsMsg'
Learn More

3-Bromo-5-(pentafluorothio)benzyl bromide, 97%, Thermo Scientific™

54.35 € - 179.00 €

Chemical Identifiers

CAS 1240257-14-0
Molecular Formula C7H5Br2F5S
Molecular Weight (g/mol) 375.977
MDL Number MFCD16652484
InChI Key PSRNYAZILJJJGP-UHFFFAOYSA-N
Synonym 3-bromo-5-pentafluorosulfur benzyl bromide, 3-bromo-5-pentafluorothio benzyl bromide, 1-bromo-3-bromomethyl-5-pentafluoro-??-sulfanyl benzeneShow MoreShow Less
PubChem CID 66523602
IUPAC Name [3-bromo-5-(bromomethyl)phenyl]-pentafluoro-$l^{6}-sulfane
SMILES C1=C(C=C(C=C1S(F)(F)(F)(F)F)Br)CBr
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15488865
SDS View Documents
Thermo Scientific Alfa Aesar
H34296.MD
250 mg N/A N/A N/A
15417237
SDS View Documents
Thermo Scientific Alfa Aesar
H34296.03
1 g N/A N/A N/A
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Specifications
Show More
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P501c

H314-H335

missing translation for 'dotInformation' : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

missing translation for 'tsca' : No

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Certificates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromo-5-(pentafluorothio)benzyl bromide, 97%, Thermo Scientific™

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.