missing translation for 'onlineSavingsMsg'
Learn More

3-Bromoadamantane-1-carboxylic acid, 97%, Thermo Scientific™

Product Code. 10394622
Click to view available options
Quantity:
1 g
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1g
This item is not returnable. View return policy

Product Code. 10394622

Brand: Thermo Scientific Maybridge NRB01917EA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 21816-08-0
Molecular Formula C11H15BrO2
Molecular Weight (g/mol) 259.143
MDL Number MFCD00167820
InChI Key DJUDQBVINJIMFO-UHFFFAOYSA-N
Synonym 1-bromo-3-adamantanecarboxylic acid, 3-bromo-1-adamantanecarboxylic acid, tricyclo 3.3.1.13,7 decane-1-carboxylic acid, 3-bromo, 3-bromo-adamantane-1-carboxylic acid, 3-adamantanecarboxylic acid, 1-bromo, 3-bromoadamantanecarboxylic acid, 3-bromotricyclo 3.3.1.1∼3,7∼ decane-1-carboxylic acid, acmc-1cnbw, maybridge1_005590, 5s,7r-3-bromoadamantane-1-carboxylate
PubChem CID 30818
IUPAC Name 3-bromoadamantane-1-carboxylic acid
SMILES C1C2CC3(CC1CC(C2)(C3)Br)C(=O)O

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 10 g
Formula Weight 259.14
Percent Purity 97%
Chemical Name or Material 3-Bromoadamantane-1-carboxylic acid
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.