missing translation for 'onlineSavingsMsg'
Learn More
3-Bromophenylacetic acid, 98+%, Thermo Scientific Chemicals Product Code.: 11475626

3-Bromophenylacetic acid, 98+%, Thermo Scientific Chemicals

Product Code. 11475626
5 g
Click to view available options
Quantity:
5 g
25 g
This item is not returnable. View return policy

Product Code. 11475626

Brand: Thermo Scientific Alfa Aesar B20758.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1878-67-7
Molecular Formula C8H7BrO2
Molecular Weight (g/mol) 215.05
MDL Number MFCD00004330
InChI Key KYNNBXCGXUOREX-UHFFFAOYSA-N
Synonym 3-bromophenylacetic acid, 2-3-bromophenyl acetic acid, 3-bromophenyl acetic acid, m-bromophenylacetic acid, benzeneacetic acid, 3-bromo, 3-bromophenylaceticacid, 3-bromo-phenylacetic acid, 3-bromphenylacetic acid, pubchem4244Show MoreShow Less
PubChem CID 74653
IUPAC Name 2-(3-bromophenyl)acetic acid
SMILES OC(=O)CC1=CC=CC(Br)=C1

Specifications

Melting Point 100°C to 102°C
Quantity 5 g
Beilstein 2045104
Formula Weight 215.05
Percent Purity ≥98%
Chemical Name or Material 3-Bromophenylacetic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10241833 Certificate TypeCertificate of Analysis Date11/07/2022 Catalog NumberB20758.06,B20758.14
Lot NumberC31P33 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20758.06,B20758.14
Lot Number10165556 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20758.06,B20758.14
Lot Number10143290 Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20758.06,B20758.14
Lot Number5003A04Q Certificate TypeCertificate of Analysis Date23/11/2021 Catalog NumberB20758.06,B20758.14

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Bromophenylacetic acid, 98+%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.