missing translation for 'onlineSavingsMsg'
Learn More

3-Butynyl p-toluenesulfonate, 96%

Product Code. 11327606
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
This item is not returnable. View return policy

Product Code. 11327606

Brand: Thermo Scientific Alfa Aesar L11166.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 23418-85-1 | C11H12O3S | 224.274 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 23418-85-1
Molecular Formula C11H12O3S
Molecular Weight (g/mol) 224.274
MDL Number MFCD00041687
InChI Key STOASOOVVADOKH-UHFFFAOYSA-N
Synonym 3-butynyl p-toluenesulfonate, but-3-yn-1-yl 4-methylbenzenesulfonate, 3-butynyl tosylate, 3-butynyl 4-toluenesulfonate, 3-1-butynyl-4-toluenesulphonate, 3-butynyl 4-methylbenzenesulfonate, p-toluenesulfonic acid 3-butynyl ester, but-3-ynyl tosylate, 4-tosyloxy-1-butyne, acmc-1cgfq
PubChem CID 272020
IUPAC Name but-3-ynyl 4-methylbenzenesulfonate
SMILES CC1=CC=C(C=C1)S(=O)(=O)OCCC#C

Specifications

Density 1.012
Boiling Point 128°C (0.01 mmHg)
Flash Point >100°C (212°F)
Refractive Index 1.522
Quantity 1 g
Beilstein 3304257
Formula Weight 224.28
Percent Purity 96%
Chemical Name or Material 3-Butynyl p-toluenesulfonate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.