missing translation for 'onlineSavingsMsg'
Learn More
Learn More
3-Chloro-2-fluoro-6-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™
Click to view available options
Quantity:
1 g
5 g
Descripción
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Especificaciones
Especificaciones
Refractive Index | 1.475 |
Quantity | 5 g |
UN Number | UN3265 |
Sensitivity | Moisture sensitive |
Formula Weight | 261 |
Percent Purity | 97% |
Chemical Name or Material | 3-Chloro-2-fluoro-6-(trifluoromethyl)benzoyl chloride |
Certificates
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
1 results found
Lot Number | Certificate Type | Date | Catalog Number |
---|---|---|---|
10141006 | Certificate of Analysis | 07/12/2021 |
Corrección del contenido de un producto
Proporcione sus comentarios sobre el contenido del producto rellenando el siguiente formulario.
Título del producto
3-Chloro-2-fluoro-6-(trifluoromethyl)benzoyl chloride, 97%, Thermo Scientific™ >
¿Detecta una oportunidad de mejora?Comparta una corrección de contenido