missing translation for 'onlineSavingsMsg'
Learn More
3-Chloro-2-fluorobenzenesulfonyl chloride, 97%, Thermo Scientific™ Product Code.: 11338876

3-Chloro-2-fluorobenzenesulfonyl chloride, 97%, Thermo Scientific™

Product Code. 11338876
1 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11338876

Brand: Thermo Scientific Alfa Aesar L19511.03

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is employed as a pharmaceutical intermediate.

Solubility
Hydrolyzes in water.

Notes
Moisture sensitive. Store in dry condition. Keep oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 351003-48-0
Molecular Formula C6H3Cl2FO2S
Molecular Weight (g/mol) 229.046
MDL Number MFCD03094234
InChI Key LIRQNYQIFFLGIE-UHFFFAOYSA-N
Synonym 3-chloro-2-fluorobenzenesulphonyl chloride, 3-chloro-2-fluorobenzene-1-sulfonyl chloride, 3-chloro-2-fluorobenzenesulphonylchloride, 3-chloro-2-fluorophenylsulfonyl chloride, 3-chloro-2-fluorobenzensulphonyl chloride, benzenesulfonyl chloride, 3-chloro-2-fluoro, pubchem8446, acmc-209vns, 3-chloro-2-fluoro-benzenesulfonyl chloride, ksc222a3dShow MoreShow Less
PubChem CID 2778321
IUPAC Name 3-chloro-2-fluorobenzenesulfonyl chloride
SMILES C1=CC(=C(C(=C1)Cl)F)S(=O)(=O)Cl

Specifications

Density 1.584
Flash Point >110°C (230°F)
Refractive Index 1.554
Quantity 1 g
UN Number UN3265
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes in water.
Formula Weight 229.06
Percent Purity 97%
Chemical Name or Material 3-Chloro-2-fluorobenzenesulfonyl chloride

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberX27A005 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberB27S012 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot NumberE27U007 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Chloro-2-fluorobenzenesulfonyl chloride, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.