missing translation for 'onlineSavingsMsg'
Learn More

3-Ethyltoluene, 97%

Product Code. p-7047341
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11389515

Brand: Thermo Scientific Alfa Aesar L18788.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 620-14-4 | C9H12 | 120.20 g/mol

It is used in the formation of 3-ethylsalicylic acid from 3-Ethyltoluene by Pseudomonas ovalis.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is used in the formation of 3-ethylsalicylic acid from 3-Ethyltoluene by Pseudomonas ovalis.

Solubility
Not miscible or difficult to mix in water.

Notes
Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition. Keep ignition sources away. Protect from electrostatic charges.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 620-14-4
Molecular Formula C9H12
Molecular Weight (g/mol) 120.20
MDL Number MFCD00009259
InChI Key ZLCSFXXPPANWQY-UHFFFAOYSA-N
Synonym 3-ethyltoluene, m-ethyltoluene, 1-methyl-3-ethylbenzene, benzene, 1-ethyl-3-methyl, m-ethylmethylbenzene, m-methylethylbenzene, toluene, m-ethyl, 3-methylethylbenzene, 1-ethyl-3-methyl-benzene, unii-737ptd7o7e
PubChem CID 12100
ChEBI CHEBI:77512
IUPAC Name 1-ethyl-3-methylbenzene
SMILES CCC1=CC=CC(C)=C1

Specifications

Melting Point -96°C
Density 0.865
Boiling Point 158°C to 159°C
Flash Point 38°C (100°F)
Refractive Index 1.496
Quantity 5 g
UN Number UN3295
Beilstein 1850821
Solubility Information Not miscible or difficult to mix in water.
Formula Weight 120.2
Percent Purity ≥98%
Chemical Name or Material 3-Ethyltoluene
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • 3-Ethyltoluene
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H226-Flammable liquid and vapour.
  • H304-May be fatal if swallowed and enters airways.
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P264-Wash face, hands and anyexposed skin thoroughly after handling.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P331-Do NOT induce vomiting.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : m-Ethyltoluene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.