missing translation for 'onlineSavingsMsg'
Learn More
3-Fluoro-L-tyrosine, 97%, Thermo Scientific Chemicals Product Code.: 11363736

3-Fluoro-L-tyrosine, 97%, Thermo Scientific Chemicals

Product Code. 11363736
50 mg
Click to view available options
Quantity:
50 mg
250 mg
This item is not returnable. View return policy

Product Code. 11363736

Brand: Thermo Scientific Alfa Aesar L16775.MF

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

3-Fluoro-L-tyrosine is used as pharma and intermediates.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
3-Fluoro-L-tyrosine is used as pharma and intermediates.

Solubility
Soluble in water (9.8 g/L).

Notes
Keep container tightly closed. Store away from oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7423-96-3
Molecular Formula C9H10FNO3
Molecular Weight (g/mol) 199.18
MDL Number MFCD00002607
InChI Key VIIAUOZUUGXERI-JLDDOWRYNA-N
Synonym 3-fluoro-l-tyrosine, s-2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, 3-fluorotyrosine, 3-fluorotyrosine, l, h-tyr 3-f-oh, unii-174nrg3m2x, s-3-fluorotyrosine, 3'-fluoro-l-tyrosine, 2s-2-amino-3-3-fluoro-4-hydroxyphenyl propanoic acid, 3-fluoro-tyrosineShow MoreShow Less
PubChem CID 643330
ChEBI CHEBI:46534
IUPAC Name (2S)-2-amino-3-(3-fluoro-4-hydroxyphenyl)propanoic acid
SMILES N[C@@H](CC1=CC=C(O)C(F)=C1)C(O)=O

Specifications

Melting Point 260°C to 261°C
UN Number UN2811
Beilstein 3204802
Solubility Information Soluble in water (9.8g/L).
Quantity 50 mg
Formula Weight 199.18
Optical Rotation −7° (c=1 in 1M HCl)
Percent Purity 97%
Chemical Name or Material 3-Fluoro-L-tyrosine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10239810 Certificate TypeCertificate of Analysis Date24/02/2023 Catalog NumberL16775.MD,L16775.MF
Lot Number10154743 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog NumberL16775.MD,L16775.MF
Lot NumberJ24R046 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL16775.MD,L16775.MF
Lot NumberZ08D046 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL16775.MD,L16775.MF
Lot NumberH05U053 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL16775.MD,L16775.MF

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-Fluoro-L-tyrosine, 97%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.