Learn More
3-Methoxy-N-methylbenzylamine, 97%, Thermo Scientific Chemicals
51.80 € - 169.85 €
Chemical Identifiers
CAS | 41789-95-1 |
---|---|
Molecular Formula | C9H14NO |
Molecular Weight (g/mol) | 152.22 |
MDL Number | MFCD04618244 |
InChI Key | FIFKRPFWLHBMHL-UHFFFAOYSA-O |
Synonym | 3-methoxy-n-methylbenzylamine, 1-3-methoxyphenyl-n-methylmethanamine, 3-methoxyphenyl methyl methyl amine, n-methyl-3-methoxybenzylamine, 3-methoxyphenyl methyl methylamine, 1-3-methoxyphenyl-n-methyl-methanamine, methyl 3-methoxybenzyl amine, ksc494s3f, n-methyl-3-methoxy-benzylamine, 3-methoxy-benzyl-methyl-amine |
PubChem CID | 5152225 |
IUPAC Name | 1-(3-methoxyphenyl)-N-methylmethanamine |
SMILES | C[NH2+]CC1=CC=CC(OC)=C1 |
Description
3-Methoxy-N-methylbenzylamine is used as an intermediate in the synthesis of diclofensine hydrochloride.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications3-Methoxy-N-methylbenzylamine is used as an intermediate in the synthesis of diclofensine hydrochloride.
Solubility
Soluble in chloroform, dichloromethane, ethyl acetate and methanol.
Notes
Keep container tightly sealed. Store in cool, dry conditions in well sealed containers. Incompatible with oxidizing agents. It is sensitive to air.
Chemical Identifiers
41789-95-1 | |
152.22 | |
FIFKRPFWLHBMHL-UHFFFAOYSA-O | |
5152225 | |
C[NH2+]CC1=CC=CC(OC)=C1 |
C9H14NO | |
MFCD04618244 | |
3-methoxy-n-methylbenzylamine, 1-3-methoxyphenyl-n-methylmethanamine, 3-methoxyphenyl methyl methyl amine, n-methyl-3-methoxybenzylamine, 3-methoxyphenyl methyl methylamine, 1-3-methoxyphenyl-n-methyl-methanamine, methyl 3-methoxybenzyl amine, ksc494s3f, n-methyl-3-methoxy-benzylamine, 3-methoxy-benzyl-methyl-amine | |
1-(3-methoxyphenyl)-N-methylmethanamine |
Specifications
41789-95-1 | |
130°C to 133°C | |
C9H14NO | |
MFCD04618244 | |
3-methoxy-n-methylbenzylamine, 1-3-methoxyphenyl-n-methylmethanamine, 3-methoxyphenyl methyl methyl amine, n-methyl-3-methoxybenzylamine, 3-methoxyphenyl methyl methylamine, 1-3-methoxyphenyl-n-methyl-methanamine, methyl 3-methoxybenzyl amine, ksc494s3f, n-methyl-3-methoxy-benzylamine, 3-methoxy-benzyl-methyl-amine | |
FIFKRPFWLHBMHL-UHFFFAOYSA-O | |
1-(3-methoxyphenyl)-N-methylmethanamine | |
5152225 | |
97% |
1.014 | |
100°C (212°F) | |
1.529 | |
Air Sensitive | |
Soluble in chloroform,dichloromethane,ethyl acetate and methanol. | |
C[NH2+]CC1=CC=CC(OC)=C1 | |
152.22 | |
151.21 | |
3-Methoxy-N-methylbenzylamine |
Safety and Handling
GHS H Statement
H301-H318-H317
Toxic if swallowed.
Causes serious eye damage.
May cause an allergic skin reaction.
P261-P264b-P270-P280-P285-P301+P312-P304+P341-P305+P351+P338-P310-P330-P342+P311-P501c
H302-H318-H334
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures; Store under Nitrogen
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.