missing translation for 'onlineSavingsMsg'
Learn More

3-(Pyrrolidin-1-ylmethyl)benzoic acid hydrochloride, 90%, Thermo Scientific™

Product Code. p-4511363
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10071974

Brand: Thermo Scientific Maybridge CC55751CB

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 887922-93-2
Molecular Formula C12H16ClNO2
Molecular Weight (g/mol) 241.715
InChI Key BORITTZEFSYNFT-UHFFFAOYSA-N
Synonym 3-pyrrolidin-1-ylmethyl benzoic acid hydrochloride, 3-1-pyrrolidylmethyl benzoic acid hydrochloride, 3-pyrrolidinylmethyl benzoic acid, chloride, 3-1-pyrrolidylmethyl benzoicacidhydrochloride, 3-pyrrolidin-1-ylmethyl-benzoic acid hydochloride, 3-pyrrolidin-1-ylmethyl-benzoic acid hydrochloride, 3-pyrrolidin-1-yl methyl benzoic acid-hydrogen chloride 1/1
PubChem CID 18525874
IUPAC Name 3-(pyrrolidin-1-ylmethyl)benzoic acid;hydrochloride
SMILES C1CCN(C1)CC2=CC=CC(=C2)C(=O)O.Cl

Specifications

Assay Percent Range 90%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 241.72
Percent Purity 90%
Chemical Name or Material 3-(Pyrrolidin-1-ylmethyl)benzoic acid hydrochloride
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.