missing translation for 'onlineSavingsMsg'
Learn More
3-(Trifluoromethoxy)benzonitrile, 97%, Thermo Scientific™ Product Code.: 11470451

3-(Trifluoromethoxy)benzonitrile, 97%, Thermo Scientific™

Product Code. 11470451
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11470451

Brand: Thermo Scientific Alfa Aesar B20007.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 52771-22-9
Molecular Formula C8H4F3NO
Molecular Weight (g/mol) 187.12
MDL Number MFCD00040959
InChI Key DCZAPXGEZYVQNX-UHFFFAOYSA-N
Synonym 3-trifluoromethoxy benzonitrile, m-trifluoromethoxybenzonitrile, benzonitrile, 3-trifluoromethoxy, 3-trifluoromethoxy-benzonitrile, 3-trifluoromethoxy benzenecarbonitrile, pubchem4825, acmc-1art8, ksc493e1j, m-trifluoromethoxy benzonitrileShow MoreShow Less
PubChem CID 142962
IUPAC Name 3-(trifluoromethoxy)benzonitrile
SMILES FC(F)(F)OC1=CC=CC(=C1)C#N

Specifications

Refractive Index 1.45
Quantity 5 g
UN Number UN3276
Beilstein 7201089
Formula Weight 187.12
Percent Purity 97%
Chemical Name or Material 3-(Trifluoromethoxy)benzonitrile

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

3 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberJ1134A Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10204929 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot NumberE3323B Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
3-(Trifluoromethoxy)benzonitrile, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.