Learn More
ibidi 3 Well Chamber, Removable
Click to view available options
:
15
Description
- Chamber slide with a removable 3 well silicone chamber
- For cell culture and immunofluorescence staining
- For use with upright and inverted microscopes
- Bottom material: microscopy glass
- Sterilized
Specifications
Specifications
| No. per Pack | 15 |