Learn More
Uvex™ 3B Chem Light Chemical Protection Coveralls
missing translation for 'orderingAttributeHoverText'
Size:
Small
Medium
Large
X-Large
2X-Large
3X-Large
Descripción
- Light and highly flexible material for high wearer comfort
- Liquid-tight
- Low-noise material
- Free from paint wetting impairment substances and silicone
- Ideal for cleaning work
- Middle finger loops for securing the sleeves
- Optimal protection thanks to self-adhesive zipper flap
- Anti-static properties
- Protection against infectious diseases
- Lined with skin-friendly non-woven material
- Secure and comfortable closures thanks to elasticated bands on hood, arms and legs
- Elasticated waistband for optimal fit
- Two-way zip
Especificaciones
Especificaciones
Product Type | Chemical Protective Clothing |
Size | 3X-Large |
Gender | Unisex |
Material | Polypropylene |
Color | White, Yellow, Orange |
Disposable | Single-use |
Waist Style | Elastic |
Ankle Style | Elastic |
Cuff Style | Covered Elastic |
Neck Style | Hooded |
Mostrar más |