missing translation for 'onlineSavingsMsg'
Learn More

(3R-cis)-(-)-3-Isopropyl-7a-methyltetrahydropyrrolo-[2,1-b]oxazol-5(6H)-one, 97%, Thermo Scientific™

Product Code. 10215831 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
250 mg
Packaging:
Glass bottle
Unit Size:
250mg
This item is not returnable. View return policy

Product Code. 10215831

Brand: Thermo Scientific Acros 326492500

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

CAS 123808-97-9
CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Boiling Point 82°C (0.7 mmHg)
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Molecular Formula C10H17NO2
Refractive Index 1.4720 to 1.474
MDL Number MFCD00269679
Quantity 250 mg
Specific Rotation Condition −93° (20°C c=4,EtOH)
Specific Rotation -93
Synonym 3r,7as-3-isopropyl-7a-methyltetrahydropyrrolo 2,1-b oxazol-5 6h-one, 3r,7as-7a-methyl-3-propan-2-yl-2,3,6,7-tetrahydropyrrolo 2,1-b 1,3 oxazol-5-one, 3r-cis---3-isopropyl-7a-methyltetrahydropyrrolo 2,1-b oxazol-5 6h-one, 3r,7as-3-isopropyl-7a-methyl-tetrahydropyrrolo 2,1-b 1,3 oxazol-5-one, 3r,7as-7a-methyl-3-propan-2-yl tetrahydropyrrolo 2,1-b 1,3 oxazol-5 6h-one, 3r,7as-7a-methyl-3-propan-2-yl-2,3,6,7-tetrahydropyrrolo 2,1-b oxazol-5-one
Solubility Information Solubility in water: .. Other solubilities: soluble in dichloromethane,chloroform
InChI Key NUUDVADIQSLTCN-WPRPVWTQSA-N
SMILES CC(C)C1COC2(N1C(=O)CC2)C
IUPAC Name (3R,7aS)-7a-methyl-3-propan-2-yl-2,3,6,7-tetrahydropyrrolo[2,1-b][1,3]oxazol-5-one
Molecular Weight (g/mol) 183.25
PubChem CID 928831
Formula Weight 183.25
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material (3R-cis)-(-)-3-Isopropyl-7a-methyltetrahydropyrrolo-[2, 1-b]oxazol-5(6H)-one, 97%
Show More Show Less
compliance-icons
Product Identifier
  • (3R-cis)-(-)-3-Isopropyl-7a-methyltetrahydropyrrolo-¢2,1-b!oxazol-5(6H)-one
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.