missing translation for 'onlineSavingsMsg'
Learn More
4-(1H-Tetrazol-5-yl)aniline, 97%, Thermo Scientific™ Product Code.: 15473657

4-(1H-Tetrazol-5-yl)aniline, 97%, Thermo Scientific™

Product Code. 15473657
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15473657

Brand: Thermo Scientific Alfa Aesar H27311.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy
Applications
Used to prepare dyes. Industrially used in natural gas and refinery process streams. They also are used in polymer industry as monomers. Amines find use as additives for lubricating oils, engine fuels, and asphalt, and are producers of ethanolamines.

Solubility
Slightly soluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 46047-18-1
Molecular Formula C7H7N5
Molecular Weight (g/mol) 161.168
MDL Number MFCD01219548
InChI Key KTPONJJKCBOJCQ-UHFFFAOYSA-N
Synonym 4-2h-tetrazol-5-yl aniline, 4-1h-tetrazol-5-yl aniline, 4-1h-tetrazol-5-yl-phenylamine, 4-1h-1,2,3,4-tetrazol-5-yl aniline, 4-2h-1,2,3,4-tetrazol-5-yl aniline, 4-2h-tetrazol-5-yl-phenylamine, 5-4-aminophenyl tetrazole, benzenamine, 4-1h-tetrazol-5-yl, benzenamine, 4-2h-tetrazol-5-yl, 5-4-aminophenyl-1h-tetrazole
PubChem CID 1079216
IUPAC Name 4-(2H-tetrazol-5-yl)aniline
SMILES C1=CC(=CC=C1C2=NNN=N2)N

Specifications

Melting Point ∽265°C (decomposition)
Quantity 250 mg
Solubility Information Slightly soluble in water.
Formula Weight 161.17
Percent Purity 97%
Chemical Name or Material 4-(1H-Tetrazol-5-yl)aniline

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10148555 Certificate TypeCertificate of Analysis Date08/12/2021 Catalog Number
Lot Number10151624 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog Number
Lot Number10174676 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10155840 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number
Lot Number10180857 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog Number

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(1H-Tetrazol-5-yl)aniline, 97%, Thermo Scientific™ >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.