missing translation for 'onlineSavingsMsg'
Learn More

AEBSF Hydrochloride, 4-(2-Aminoethyl)-benzenesulfonyl Fluoride HCl, White Powder, Fisher BioReagents

37.75 € - 823.00 €

Chemical Identifiers

CAS 30827-99-7
Molecular Formula C8H11ClFNO2S
Molecular Weight (g/mol) 239.689
InChI Key WRDABNWSWOHGMS-UHFFFAOYSA-N
Synonym 4-2-aminoethyl benzenesulfonyl fluoride hydrochloride, aebsf, aebsf hydrochloride, pefabloc sc, aebsf hcl, pefabloc, 4-2-aminoethyl benzene-1-sulfonyl fluoride hydrochloride, aebsf, hydrochloride, benzenesulfonyl fluoride, 4-2-aminoethyl-, hydrochloride, 4-2-aminoethyl benzenesulfonylfluoride hydrochloride
PubChem CID 186136
IUPAC Name 4-(2-aminoethyl)benzenesulfonyl fluoride;hydrochloride
SMILES C1=CC(=CC=C1CCN)S(=O)(=O)F.Cl
View More Specs

Products 5
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
10357924
SDS View Documents
Fisher BioReagents
BP264410
10 mg Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10707735
SDS View Documents
Fisher BioReagents
BP2644-50
50 mg Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12194471
SDS View Documents
Fisher BioReagents
BP2644-100
100 mg Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
10104144
SDS View Documents
Fisher BioReagents
BP2644-500
500 mg Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
11747499
SDS View Documents
Fisher BioReagents
BP2644-1
1 g Amber Glass This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
Description

Description

  • Irreverisble serine protease inhibitor
  • Inhibits proteases like trypsin, chymotrypsin, kallikrein, plasmin, and thrombin
  • Acts as Sulfonyl fluoride that reacts with hydroxy group of the active site serine residue
  • More soluble and less toxic
Specifications
Show More
Videos
Safety and Handling

Safety and Handling

Emergency Overview
Corrosive. Irritating to eyes and skin. May cause burns.
NFPA
Health: 3
Flammability:

Signal Word: DANGER!

missing translation for 'tsca' : Not on TSCA inventory: for R and D use only; not for manufacturing or commercial purposes.

Recommended Storage : RT, protect from light.

SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
AEBSF Hydrochloride, 4-(2-Aminoethyl)-benzenesulfonyl Fluoride HCl, White Powder, Fisher BioReagents

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.