missing translation for 'onlineSavingsMsg'
Learn More
4-(2-Hydroxyethyl)benzonitrile, 98%, Thermo Scientific Chemicals Product Code.: 11319677

4-(2-Hydroxyethyl)benzonitrile, 98%, Thermo Scientific Chemicals

Product Code. 11319677
250 mg
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 11319677

Brand: Thermo Scientific Alfa Aesar 041283.MD

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Organic intermediate

4-(2-Hydroxyethyl)benzonitrile is used to produce 4-(2-bromo-ethyl)-benzonitrile. It is used as organic intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69395-13-7
Molecular Formula C9H9NO
Molecular Weight (g/mol) 147.177
MDL Number MFCD00052461
InChI Key RBSJBNYPTGMZIH-UHFFFAOYSA-N
Synonym 4-2-hydroxyethyl benzonitrile, benzonitrile, 4-2-hydroxyethyl, 2-4-cyanophenyl ethanol, 2-4-cyanophenyl ethan-1-one, 4-2-hydroxyethyl benzenecarbonitrile, 4-cyanobenzeneethanol, p-cyanophenethylalcohol, p-cyanophenethyl alcohol, 4-cyanophenethyl alcohol, acmc-20a4x6Show MoreShow Less
PubChem CID 2800815
IUPAC Name 4-(2-hydroxyethyl)benzonitrile
SMILES C1=CC(=CC=C1CCO)C#N

Specifications

Boiling Point 116°C to 118°C (0.5 mmHg)
Quantity 250 mg
UN Number UN3439
Formula Weight 147.18
Percent Purity 98%
Physical Form Chunks and/or Powder
Chemical Name or Material 4-(2-Hydroxyethyl)benzonitrile

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(2-Hydroxyethyl)benzonitrile, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.