Learn More
Memmert™ 4-20 mA Current Loop Interface for UF750
Temperature Output for external temperature monitoring.
Brand: Memmert™ UF750V6
This item is not returnable.
View return policy
Description
- Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring (max. 1 SingleDISPLAY, max. 3 TwinDISPLAY)
Specifications
UF750 Oven | |
Current Loop Interface | |
4 to 20 mA |
Product Suggestions
Customers who viewed this item also viewed.
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ 4-20 mA Current Loop Interface for UF750 > For Use With: UF750
Spot an opportunity for improvement?Share a Content Correction