Learn More
Memmert™ 4-20 mA Current Loop Interface for UN260plus
Temperature Output for external temperature monitoring.
Brand:  Memmert™ UN260plusV6
Product Code. 17560046
  • 451.00 € / Each
Estimated Shipment: 21-05-2025
to see stock.
Explore more special offers
Add to basket
This item is not returnable. View return policy
Description
Description
  • Temperature of a Pt100 sensor positioned flexibly in chamber for external temperature monitoring (max. 1 SingleDISPLAY, max. 3 TwinDISPLAY)
Specifications
Specifications
UN260plus Oven
Current Loop Interface
4 to 20 mA
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Memmert™ 4-20 mA Current Loop Interface for UN260plus > For Use With: UN260plus