Learn More
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine, 97%, Thermo Scientific Chemicals
117.59 € - 370.85 €
Chemical Identifiers
CAS | 181219-01-2 |
---|---|
Molecular Formula | C11H16BNO2 |
Molecular Weight (g/mol) | 205.06 |
MDL Number | MFCD01319051 |
InChI Key | NLTIETZTDSJANS-UHFFFAOYSA-N |
Synonym | 4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 4-pyridineboronic acid pinacol ester, pyridine-4-boronic acid pinacol ester, 4-pyridylboronic acid pinacol ester, pyridine-4-boronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine-4-boronic pinacolate, pyridine-4-boronic acid pinacol cyclic ester, 2-4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, pyridine-4-boronic pinacol esterShow More |
PubChem CID | 2734648 |
IUPAC Name | 4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
SMILES | CC1(C)OB(OC1(C)C)C1=CC=NC=C1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10202382
|
Thermo Scientific Acros
367510010 |
1 g | Glass bottle |
117.59 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10552992
|
Thermo Scientific Acros
367510050 |
5 g | Glass bottle |
370.85 €
5g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
181219-01-2 | |
205.06 | |
NLTIETZTDSJANS-UHFFFAOYSA-N | |
2734648 | |
CC1(C)OB(OC1(C)C)C1=CC=NC=C1 |
C11H16BNO2 | |
MFCD01319051 | |
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 4-pyridineboronic acid pinacol ester, pyridine-4-boronic acid pinacol ester, 4-pyridylboronic acid pinacol ester, pyridine-4-boronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine-4-boronic pinacolate, pyridine-4-boronic acid pinacol cyclic ester, 2-4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, pyridine-4-boronic pinacol esterShow More | |
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
Specifications
181219-01-2 | |
100.0 | |
White | |
96% min. (GC) | |
MFCD01319051 | |
Solubility in water: insoluble | |
CC1(C)OB(OC1(C)C)C1=CC=NC=C1 | |
205.06 | |
205.06 | |
Powder |
96.0 | |
153.0°C to 155.0°C | |
Authentic | |
C11H16BNO2 | |
4-4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, 4-pyridineboronic acid pinacol ester, pyridine-4-boronic acid pinacol ester, 4-pyridylboronic acid pinacol ester, pyridine-4-boronic acid, pinacol ester, 4-tetramethyl-1,3,2-dioxaborolan-2-yl pyridine, pyridine-4-boronic pinacolate, pyridine-4-boronic acid pinacol cyclic ester, 2-4-pyridyl-4,4,5,5-tetramethyl-1,3,2-dioxaborolane, pyridine-4-boronic pinacol ester | |
NLTIETZTDSJANS-UHFFFAOYSA-N | |
4-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine | |
2734648 | |
97% | |
4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine |
Safety and Handling
GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS Signal Word: Warning
Recommended Storage : Refrigerator +4°C
- 4-(4,4,5,5-Tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.