missing translation for 'onlineSavingsMsg'
Learn More
4,4-Dimethyl-2-oxazoline, 98%, Thermo Scientific Chemicals Product Code.: 11400553

4,4-Dimethyl-2-oxazoline, 98%, Thermo Scientific Chemicals

Product Code. 11400553
100 g
Click to view available options
Quantity:
5 g
25 g
100 g
This item is not returnable. View return policy

Product Code. 11400553

Brand: Thermo Scientific Alfa Aesar A12670.22

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

4,4-Dimethyl-2-oxazoline is used in the fictionalization of a non-activated carbon-hydrogen bond.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4,4-Dimethyl-2-oxazoline is used in the fictionalization of a non-activated carbon-hydrogen bond.

Solubility
Hydrolyzes with water.

Notes
Moisture Sensitive. Store in cool dry place in tightly closed container. With good ventilation. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 30093-99-3
Molecular Formula C5H9NO
Molecular Weight (g/mol) 99.13
MDL Number MFCD00014507
InChI Key KOAMXHRRVFDWRQ-UHFFFAOYSA-N
Synonym 4,4-dimethyl-2-oxazoline, 4,4-dimethyloxazoline, 4,4-dimethyl-4,5-dihydrooxazole, oxazole, 4,5-dihydro-4,4-dimethyl, 4,5-dihydro-4,4-dimethyloxazole, pubchem19647, acmc-1cjzq, koamxhrrvfdwrq-uhfffaoysa, 4,4-dimethyl-4,5-dihydro-1,3-oxazole, 4,4-dimethyl-5h-1,3-oxazol-5-ylShow MoreShow Less
PubChem CID 121630
IUPAC Name 4,4-dimethyl-5H-1,3-oxazole
SMILES CC1(C)COC=N1

Specifications

Density 0.94
Boiling Point 99°C to 100°C
Flash Point 1°C (33°F)
Refractive Index 1.419
Quantity 100 g
UN Number UN1993
Beilstein 1209606
Sensitivity Moisture sensitive
Solubility Information Hydrolyzes with water.
Formula Weight 99.13
Percent Purity 98%
Chemical Name or Material 4,4-Dimethyl-2-oxazoline
Show More Show Less

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10249332 Certificate TypeCertificate of Analysis Date12/01/2024 Catalog NumberA12670.14,A12670.06,A12670.22
Lot Number10244883 Certificate TypeCertificate of Analysis Date01/12/2022 Catalog NumberA12670.14,A12670.06,A12670.22
Lot Number10244659 Certificate TypeCertificate of Analysis Date16/11/2022 Catalog NumberA12670.14,A12670.06,A12670.22
Lot Number10181252 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12670.14,A12670.06,A12670.22
Lot Number10179759 Certificate TypeCertificate of Analysis Date22/11/2021 Catalog NumberA12670.14,A12670.06,A12670.22
compliance-icons
Product Identifier
  • 4,4-Dimethyl-2-oxazoline
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 2
Hazard Statement
  • H225-Highly flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P240-Ground/bond container and receiving equipment.
Supplemental information
  • MIXTURE LIST-Contains : 4,5-Dihydro-4,4-dimethyloxazole

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4,4-Dimethyl-2-oxazoline, 98%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.