Learn More
4-(5,6-Dihydro-4H-1,3-oxazin-2-yl)benzeneboronic acid, 96%, Thermo Scientific Chemicals Product Code.: 15460346
4-(5,6-Dihydro-4H-1,3-oxazin-2-yl)benzeneboronic acid, 96%, Thermo Scientific Chemicals
Product Code. 15460346
250 mg
Click to view available options
Quantity:
250 mg
1 g
Product Code. 15460346
Brand: Thermo Scientific Alfa Aesar H53062.MD
Request Bulk or Custom Format
Please to purchase this item. Need a web account? Register with us today!
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 850568-68-2
Molecular Formula C10H12BNO3
Molecular Weight (g/mol) 205.02
MDL Number MFCD06659833
InChI Key OASSAUSIPZASKK-UHFFFAOYSA-N
Synonym 4-5,6-dihydro-4h-1,3-oxazin-2-yl phenyl boronic acid, 2-4-boronophenyl-5,6-dihydro-4h-1,3-oxazine, 2-4-boronobenzene-5,6-dihydro-4h-1,3-oxazine, 4-5,6-dihydro-4h-1,3-oxazin-2-yl phenylboronic acid, 4-5,6-dihydro-4h-1,3-oxazin-2-yl benzeneboronic acid, boronic acid,b-4-5,6-dihydro-4h-1,3-oxazin-2-yl phenyl
PubChem CID 44119325
IUPAC Name [4-(5,6-dihydro-4H-1,3-oxazin-2-yl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)C2=NCCCO2)(O)O
Specifications
Melting Point 180°C to 182°C
Quantity 250 mg
Formula Weight 205.02
Percent Purity 96%
Chemical Name or Material 4-(5,6-Dihydro-4H-1,3-oxazin-2-yl)benzeneboronic acid
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
4-(5,6-Dihydro-4H-1,3-oxazin-2-yl)benzeneboronic acid, 96%, Thermo Scientific Chemicals >