Learn More
4-[(6-Methylpyrazin-2-yl)oxy]benzenesulfonyl chloride, 97%, Thermo Scientific™
124.00 € - 383.00 €
Chemical Identifiers
CAS | 926921-66-6 |
---|---|
Molecular Formula | C11H9ClN2O3S |
Molecular Weight (g/mol) | 284.71 |
MDL Number | MFCD09879936 |
InChI Key | GUJMBSPJUZJPEB-UHFFFAOYSA-N |
Synonym | 4-6-methylpyrazin-2-yl oxy benzenesulfonyl chloride, 4-6-methylpyrazin-2-yl oxy benzene-1-sulfonyl chloride |
PubChem CID | 24229688 |
IUPAC Name | 4-(6-methylpyrazin-2-yl)oxybenzenesulfonyl chloride |
SMILES | CC1=CN=CC(OC2=CC=C(C=C2)S(Cl)(=O)=O)=N1 |
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Packaging | Price | Quantity & Availability | ||||
10347444
|
Thermo Scientific Maybridge
CC62703CB |
250 mg | Amber Glass Bottle |
124.00 €
250mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
10718265
|
Thermo Scientific Maybridge
CC62703DA |
1 g | Amber Glass Bottle |
383.00 €
1g |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Chemical Identifiers
926921-66-6 | |
284.71 | |
GUJMBSPJUZJPEB-UHFFFAOYSA-N | |
24229688 | |
CC1=CN=CC(OC2=CC=C(C=C2)S(Cl)(=O)=O)=N1 |
C11H9ClN2O3S | |
MFCD09879936 | |
4-6-methylpyrazin-2-yl oxy benzenesulfonyl chloride, 4-6-methylpyrazin-2-yl oxy benzene-1-sulfonyl chlorideShow More | |
4-(6-methylpyrazin-2-yl)oxybenzenesulfonyl chloride |
Specifications
926921-66-6 | |
100.0 | |
C11H9ClN2O3S | |
4-6-methylpyrazin-2-yl oxy benzenesulfonyl chloride, 4-6-methylpyrazin-2-yl oxy benzene-1-sulfonyl chloride | |
CC1=CN=CC(OC2=CC=C(C=C2)S(Cl)(=O)=O)=N1 | |
284.71 | |
284.72 | |
4-[(6-Methylpyrazin-2-yl)oxy]benzenesulfonyl chloride |
96.5 | |
97% | |
MFCD09879936 | |
GUJMBSPJUZJPEB-UHFFFAOYSA-N | |
4-(6-methylpyrazin-2-yl)oxybenzenesulfonyl chloride | |
24229688 | |
97% |
Safety and Handling
GHS H Statement:
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Contact with water liberates toxic gas.
Reacts violently with water.
GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
Your input is important to us. Please complete this form to provide feedback related to the content on this product.