missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-2-chlorotoluene, 98%, Thermo Scientific Chemicals

Product Code. p-7026339
Click to view available options
Quantity:
25 g
100 g
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 11475966

Brand: Thermo Scientific Alfa Aesar B22293.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 89794-02-5
Molecular Formula C7H6BrCl
Molecular Weight (g/mol) 205.479
MDL Number MFCD00060651
InChI Key LIFMTDJMLRECMX-UHFFFAOYSA-N
Synonym 4-bromo-2-chlorotoluene, 2-chloro-4-bromotoluene, 4-bromo-2-chloro toluene, 4-bromo-2-chloro-1-methyl-benzene, benzene, 4-bromo-2-chloro-1-methyl, 4-bromo-chlorotoluene, pubchem3603, 4-bromo-2chlorotoluene, 2-chloro 4-bromo toluene, acmc-209r2x
PubChem CID 2735554
IUPAC Name 4-bromo-2-chloro-1-methylbenzene
SMILES CC1=C(C=C(C=C1)Br)Cl

Specifications

Density 1.538
Boiling Point 212°C
Quantity 25 g
Refractive Index 1.5735
Beilstein 3234853
Formula Weight 205.48
Percent Purity 98%
Chemical Name or Material 4-Bromo-2-chlorotoluene

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.