missing translation for 'onlineSavingsMsg'
Learn More

4-Bromo-N-(4-fluorophenyl)benzamide, 97%, Thermo Scientific™

Product Code. 15453878
Click to view available options
Quantity:
250 mg
1 g
This item is not returnable. View return policy
This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 346723-41-9
Molecular Formula C13H9BrFNO
Molecular Weight (g/mol) 294.123
MDL Number MFCD00442009
InChI Key DBCKBDOQSKIIOA-UHFFFAOYSA-N
Synonym 4-bromo-n-4-fluorophenyl benzamide, benzamide, n-4-fluorophenyl-4-bromo, cambridge id 5270654, n-4-fluorophenyl-4-bromobenzamide, 4-bromo-n-4-fluoro-phenyl-benzamide, 4-bromophenyl-n-4-fluorophenyl carboxamide
PubChem CID 532150
IUPAC Name 4-bromo-N-(4-fluorophenyl)benzamide
SMILES C1=CC(=CC=C1C(=O)NC2=CC=C(C=C2)F)Br

Specifications

Quantity 1 g
Formula Weight 294.12
Percent Purity 97%
Chemical Name or Material 4-Bromo-N-(4-fluorophenyl)benzamide
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'