missing translation for 'onlineSavingsMsg'
Learn More
4-Chloro-5,6,7,8-tetrahydro-1-benzothieno[2,3-d]pyrimidine, 96%, Thermo Scientific Chemicals Product Code.: 15438065

4-Chloro-5,6,7,8-tetrahydro-1-benzothieno[2,3-d]pyrimidine, 96%, Thermo Scientific Chemicals

Product Code. 15438065
5 g
Click to view available options
Quantity:
250 mg
1 g
5 g
This item is not returnable. View return policy

Product Code. 15438065

Brand: Thermo Scientific Alfa Aesar H32353.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 40493-18-3
Molecular Formula C10H9ClN2S
Molecular Weight (g/mol) 224.71
MDL Number MFCD00463624
InChI Key PRNJDUCSVXTOTN-UHFFFAOYSA-N
Synonym 4-chloro-5,6,7,8-tetrahydrobenzo 4,5 thieno 2,3-d pyrimidine, 4-chloro-5,6,7,8-tetrahydro 1 benzothieno 2,3-d pyrimidine, 4-chloro-5,6,7,8-tetrahydro-benzo 4,5 thieno 2,3-d pyrimidine, 4-chloro-5h,6h,7h,8h-pyrimido 4,5-b benzothiophene, 1 benzothieno 2,3-d pyrimidine,4-chloro-5,6,7,8-tetrahydro, 4-chloro-5,6,7,8-tetrahydro-1 benzothiolo 2,3-d pyrimidine, 3-chloro-8-thia-4,6-diazatricyclo 7.4.0.0^ 2,7 trideca-1 9 ,2 7 ,3,5-tetraene, 4-chloro-5,6,7,8-tetrahydrobenzo b thiopheno 2,3-d pyrimidine, 3-chloro-8-thia-4,6-diazatricyclo 7.4.0.0 2 ,? trideca-1 9 ,2,4,6-tetraeneShow MoreShow Less
PubChem CID 699515
IUPAC Name 4-chloro-5,6,7,8-tetrahydro-[1]benzothiolo[2,3-d]pyrimidine
SMILES ClC1=C2C3=C(CCCC3)SC2=NC=N1

Specifications

Melting Point 95°C to 96°C
Quantity 5 g
Formula Weight 192.65
Percent Purity 96%
Chemical Name or Material 4-Chloro-5,6,7,8-tetrahydro-1-benzothieno[2,3-d]pyrimidine

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

1 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10161128 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberH32353.06,H32353.03

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-Chloro-5,6,7,8-tetrahydro-1-benzothieno[2,3-d]pyrimidine, 96%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.