missing translation for 'onlineSavingsMsg'
Learn More

4-Chlorobenzyl alcohol, 99%

Product Code. p-85667 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
25 g
100 g
Packaging:
Glass bottle
Unit Size:
100g
25g
This item is not returnable. View return policy

Product Code. 10142660

Brand: Thermo Scientific Acros undefined

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 873-76-7 | C7H7ClO | 142.58 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 873-76-7
Molecular Formula C7H7ClO
Molecular Weight (g/mol) 142.58
MDL Number MFCD00004652
InChI Key PTHGDVCPCZKZKR-UHFFFAOYSA-N
Synonym 4-chlorobenzyl alcohol, 4-chlorophenyl methanol, p-chlorobenzyl alcohol, benzenemethanol, 4-chloro, 4-chlorobenzenemethanol, 4-chlorobenzylalcohol, benzyl alcohol, p-chloro, 4-chlorobenzylic alcohol, unii-gf8bs53b6v, ccris 5120
PubChem CID 13397
IUPAC Name (4-chlorophenyl)methanol
SMILES C1=CC(=CC=C1CO)Cl

Specifications

CAS Min % 98.5
CAS Max % 100.0
Melting Point 69.0°C to 72.0°C
Color Beige to White
Boiling Point 234.0°C
Flash Point 70°C
Infrared Spectrum Authentic
Assay Percent Range 99%
Packaging Glass bottle
Linear Formula ClC6H4CH2OH
Quantity 25 g
Beilstein 06, 444
Fieser 07,59
Solubility Information Solubility in water: 2.5g/L in water (20°C)
Formula Weight 142.58
Percent Purity 99%
Physical Form Crystalline Needles or Powder
Chemical Name or Material 4-Chlorobenzyl alcohol, 99%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.