missing translation for 'onlineSavingsMsg'
Learn More
4-(Cyanomethyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals Product Code.: 11364326

4-(Cyanomethyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals

Product Code. 11364326
5 g
Click to view available options
Quantity:
1 g
5 g
This item is not returnable. View return policy

Product Code. 11364326

Brand: Thermo Scientific Alfa Aesar L19955.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 91983-26-5
Molecular Formula C8H8BNO2
Molecular Weight (g/mol) 160.967
MDL Number MFCD01632200
InChI Key YKVMTTIYBSVPEQ-UHFFFAOYSA-N
Synonym 4-cyanomethyl benzeneboronic acid, 4-cyanomethyl phenyl boronic acid, 4-cyanomethylphenylboronic acid, 4-cyanomethylphenyl boronic acid, 4-cyanomethyl phenylboronic acid, 4-boronophenylacetonitrile, boronic acid, 4-cyanomethyl phenyl, pubchem7789, acmc-209reeShow MoreShow Less
PubChem CID 2773342
IUPAC Name [4-(cyanomethyl)phenyl]boronic acid
SMILES B(C1=CC=C(C=C1)CC#N)(O)O

Specifications

Melting Point ∼196°C to 200°C
Quantity 5 g
Beilstein 3031323
Formula Weight 160.97
Percent Purity 98%
Chemical Name or Material 4-(Cyanomethyl)benzeneboronic acid

Certificates

A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.

5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number10111885 Certificate TypeCertificate of Analysis Date07/12/2021 Catalog NumberL19955.03,L19955.06
Lot Number10155856 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL19955.03,L19955.06
Lot Number10106602 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL19955.03,L19955.06
Lot Number10204660 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL19955.03,L19955.06
Lot Number10153299 Certificate TypeCertificate of Analysis Date06/12/2021 Catalog NumberL19955.03,L19955.06

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title
4-(Cyanomethyl)benzeneboronic acid, 95%, Thermo Scientific Chemicals >

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.