Learn More
4-Hydroxy-2-(trifluoromethyl)benzoic acid, 98%, Thermo Scientific™
54.40 € - 150.00 €
Chemical Identifiers
CAS | 320-32-1 |
---|---|
Molecular Formula | C8H5F3O3 |
Molecular Weight (g/mol) | 206.12 |
MDL Number | MFCD01091007 |
InChI Key | CSQAVQLFCBRQJM-UHFFFAOYSA-N |
Synonym | 4-hydroxy-2-trifluoromethyl benzoic acid, benzoic acid, 4-hydroxy-2-trifluoromethyl, pubchem13890, acmc-1ahif, 2-carboxy-5-hydroxybenzotrifluoride, 4-carboxy-3-trifluoromethyl phenol, 2-trifluoromethyl-4-hydroxybenzoic acid, 4-hydroxy-2-trifluoromethyl-benzoic acid, 4-hydroxy-2-trifluoromethyl benzoicacid |
PubChem CID | 2775111 |
IUPAC Name | 4-hydroxy-2-(trifluoromethyl)benzoic acid |
SMILES | C1=CC(=C(C=C1O)C(F)(F)F)C(=O)O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications4-Hydroxy-2-(trifluoromethyl)benzoic acid is used in pharm industry.
Solubility
Slightly soluble in water.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Chemical Identifiers
320-32-1 | |
206.12 | |
CSQAVQLFCBRQJM-UHFFFAOYSA-N | |
2775111 | |
C1=CC(=C(C=C1O)C(F)(F)F)C(=O)O |
C8H5F3O3 | |
MFCD01091007 | |
4-hydroxy-2-trifluoromethyl benzoic acid, benzoic acid, 4-hydroxy-2-trifluoromethyl, pubchem13890, acmc-1ahif, 2-carboxy-5-hydroxybenzotrifluoride, 4-carboxy-3-trifluoromethyl phenol, 2-trifluoromethyl-4-hydroxybenzoic acid, 4-hydroxy-2-trifluoromethyl-benzoic acid, 4-hydroxy-2-trifluoromethyl benzoicacid | |
4-hydroxy-2-(trifluoromethyl)benzoic acid |
Specifications
320-32-1 | |
C8H5F3O3 | |
4-hydroxy-2-trifluoromethyl benzoic acid, benzoic acid, 4-hydroxy-2-trifluoromethyl, pubchem13890, acmc-1ahif, 2-carboxy-5-hydroxybenzotrifluoride, 4-carboxy-3-trifluoromethyl phenol, 2-trifluoromethyl-4-hydroxybenzoic acid, 4-hydroxy-2-trifluoromethyl-benzoic acid, 4-hydroxy-2-trifluoromethyl benzoicacid | |
CSQAVQLFCBRQJM-UHFFFAOYSA-N | |
4-hydroxy-2-(trifluoromethyl)benzoic acid | |
2775111 | |
98% |
159°C to 161°C | |
MFCD01091007 | |
Slightly soluble in water. | |
C1=CC(=C(C=C1O)C(F)(F)F)C(=O)O | |
206.12 | |
206.12 | |
4-Hydroxy-2-(trifluoromethyl)benzoic acid |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
H315-H319-H335
missing translation for 'tsca' : No
Recommended Storage : Ambient temperatures
Your input is important to us. Please complete this form to provide feedback related to the content on this product.