missing translation for 'onlineSavingsMsg'
Learn More

4-Iodo-3-nitrobenzoic acid, 97%

Product Code. 11491647
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11491647

Brand: Thermo Scientific Alfa Aesar B25626.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 35674-27-2 | C7H4INO4 | 293.02 g/mol

4-Iodo-3-nitrobenzoic acid is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
4-Iodo-3-nitrobenzoic acid is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Light sensitive. Store away from light, oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 35674-27-2
Molecular Formula C7H4INO4
Molecular Weight (g/mol) 293.02
MDL Number MFCD00502399
InChI Key DNMTZLCNLAIKQC-UHFFFAOYSA-N
Synonym 4-iodo-3-nitrobenzoic acid, 4-iodo-3-nitrobenzoicacid, 3-nitro-4-iodobenzoic acid, 4-iodo-3-nitro-benzoic acid, benzoic acid, 4-iodo-3-nitro, pubchem13471, acmc-209z8y, 4-iodo-3-nitro benzoic acid
PubChem CID 11208598
IUPAC Name 4-iodo-3-nitrobenzoic acid
SMILES OC(=O)C1=CC(=C(I)C=C1)[N+]([O-])=O

Specifications

Melting Point 208°C to 211°C
Quantity 5 g
Sensitivity Light sensitive
Solubility Information Insoluble in water.
Formula Weight 293.02
Percent Purity 97%
Chemical Name or Material 4-Iodo-3-nitrobenzoic acid

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.